Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ASF1B Mouse anti-Human, Clone:1A6G4, Abnova™

Catalog No. 89334352 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-334-352 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-334-352 Supplier Abnova Corporation Supplier No. MAB17904
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against full length recombinant human ASF1B.

This gene encodes a member of the H3/H4 family of histone chaperone proteins and is similar to the anti-silencing function-1 gene in yeast. The encoded protein is the substrate of the tousled-like kinase family of cell cycle-regulated kinases, and may play a key role in modulating the nucleosome structure of chromatin by ensuring a constant supply of histones at sites of nucleosome assembly. [provided by RefSeq]

Specifications

Antigen ASF1B
Applications ELISA, Western Blot
Classification Monoclonal
Clone 1A6G4
Conjugate Unconjugated
Description ASF1 anti-silencing function 1 homolog B (S. cerevisiae)
Dilution ELISA (1:10000)
Western Blot (1:100-1:500)
The optimal working dilution should be determined by the end user.
Formulation In PBS (0.05% sodium azide)
Gene ASF1B
Gene Alias CIA-II, FLJ10604
Host Species Mouse
Immunogen Recombinant protein corresponding to full length human ASF1B.
Quantity 100 μg
Gene ID (Entrez) 55723
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.