Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ASF1B Monoclonal Antibody (02)
ASF1B Monoclonal Antibody (2)
Supplier: Thermo Scientific MA529040
Description
ASF1B Monoclonal Antibody for ICC/IF

Specifications
ASF1B | |
Monoclonal | |
1 mg/mL | |
PBS with no preservative | |
Q9NVP2 | |
ASF1B | |
Recombinant Human ASF1B protein (Met1-Ile202). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG2b |
Immunocytochemistry | |
02 | |
Unconjugated | |
ASF1B | |
1700003K02Rik; AA409591; anti-silencing function 1B histone chaperone; anti-silencing function protein 1 homolog B; ASF1 anti-silencing function 1 homolog B; Asf1b; CCG1-interacting factor A-II; CIA-II; hAsf1; hAsf1b; hCIA-II; histone chaperone ASF1B; mCIA-II | |
Mouse | |
Protein A | |
RUO | |
55723 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction