Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ASF1B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA534866
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA534866 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA534866 Supplier Invitrogen™ Supplier No. PA534866
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: rat testis. Predicted reactivity: Mouse (89%), Rat (90%), Pig (95%), Chimpanzee (99%), Bovine (96%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

This gene encodes a member of the H3/H4 family of histone chaperone proteins and is similar to the anti-silencing function-1 gene in yeast. The encoded protein is the substrate of the tousled-like kinase family of cell cycle-regulated kinases, and may play a key role in modulating the nucleosome structure of chromatin by ensuring a constant supply of histones at sites of nucleosome assembly.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ASF1B
Applications Western Blot
Classification Polyclonal
Concentration 0.6 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7
Gene ASF1B
Gene Accession No. Q9NVP2
Gene Alias 1700003K02Rik; AA409591; anti-silencing function 1B histone chaperone; anti-silencing function protein 1 homolog B; ASF1 anti-silencing function 1 homolog B; Asf1b; CCG1-interacting factor A-II; CIA-II; hAsf1; hAsf1b; hCIA-II; histone chaperone ASF1B; mCIA-II
Gene Symbols ASF1B
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human ASF1B. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 304648, 55723
Target Species Human, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.