Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ASGPR1 Antibody (114), Allophycocyanin, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP290400APC
Description
ASGPR1 Monoclonal antibody specifically detects ASGPR1 in Mouse samples. It is validated for Immunohistochemistry (Paraffin)Specifications
ASGPR1 | |
Monoclonal | |
APC | |
PBS | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Mouse | |
Purified |
Immunohistochemistry (Paraffin) | |
114 | |
Immunohistochemistry-Paraffin | |
ASGPR, ASGPR 1, ASGP-R 1, asialoglycoprotein receptor 1, CLEC4H1ASGPR1, C-type lectin domain family 4 member H1, Hepatic lectin H1, HL-1 | |
Rabbit immunized with purified, recombinant Mouse ASGPR1 (Accession#: NP_033844.1, Ser60-Asn284) | |
0.1 mL | |
Immunology, Virology Bacteria and Parasites | |
432 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction