Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ASGPR1 Antibody (950203), mFluor Violet 500 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB43941MFV500
Description
ASGPR1 Monoclonal antibody specifically detects ASGPR1 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, CyTOFSpecifications
| ASGPR1 | |
| Monoclonal | |
| mFluor Violet 500 SE | |
| 50mM Sodium Borate | |
| Mouse | |
| 0.1 mL | |
| Immunology, Virology Bacteria and Parasites | |
| 432 | |
| Store at 4C in the dark. |
| Flow Cytometry, Immunohistochemistry, CyTOF | |
| 950203 | |
| Flow Cytometry, Immunohistochemistry, CyTOF-ready | |
| ASGPR, ASGPR 1, ASGP-R 1, asialoglycoprotein receptor 1, CLEC4H1ASGPR1, C-type lectin domain family 4 member H1, Hepatic lectin H1, HL-1 | |
| Mouse myeloma cell line NS0-derived recombinant human ASGR1/ASGPR1, Gln62-Leu291, Accession # P07306 | |
| RUO | |
| Primary | |
| Human | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction