Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ASGPR1 Antibody (950203), PerCP, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB43941C
Description
ASGPR1 Monoclonal antibody specifically detects ASGPR1 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, CyTOFSpecifications
ASGPR1 | |
Monoclonal | |
PerCP | |
PBS | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Primary | |
Human | |
Purified |
Flow Cytometry, Immunohistochemistry, CyTOF | |
950203 | |
Flow Cytometry, Immunohistochemistry, CyTOF-ready | |
ASGPR, ASGPR 1, ASGP-R 1, asialoglycoprotein receptor 1, CLEC4H1ASGPR1, C-type lectin domain family 4 member H1, Hepatic lectin H1, HL-1 | |
Mouse myeloma cell line NS0-derived recombinant human ASGR1/ASGPR1, Gln62-Leu291, Accession # P07306 | |
0.1 mL | |
Immunology, Virology Bacteria and Parasites | |
432 | |
Store at 4°C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction