Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human ASGR1/ASGPR1 Alexa Fluor 750-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems FAB43941S100UG
This item is not returnable.
View return policy
Description
ASGPR1 Monoclonal specifically detects ASGPR1 in Human samples. It is validated for Flow Cytometry.Specifications
ASGPR1 | |
Monoclonal | |
Alexa Fluor 750 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. | |
ASGPR, ASGPR 1, ASGP-R 1, ASGPR1, asialoglycoprotein receptor 1, CLEC4H1, CLEC4H1ASGPR1, C-type lectin domain family 4 member H1, Hepatic lectin H1, HL-1, MHL1, RHL1 | |
Mouse myeloma cell line NS0-derived recombinant human ASGR1/ASGPR1, Gln62-Leu291, Accession No. P07306 | |
100 μg | |
432.0 | |
Human | |
Supernatant |
Flow Cytometry | |
950203 | |
Flow Cytometry 0.25-1 μg/106 cells | |
P07306 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
Primary | |
Detects human ASGR1/ASGPR1 in direct ELISAs. | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 °C as supplied. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction