Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ASGPR1 Rabbit anti-Human, Clone: 24, Novus Biologicals™

Rabbit Monoclonal Antibody
$353.50 - $560.50
Specifications
Antigen | ASGPR1 |
---|---|
Clone | 24 |
Dilution | Western Blot 1:20000-1:100000, Sandwich ELISA Detection 1:1000-1:10000 |
Applications | Western Blot, ELISA |
Classification | Monoclonal |
Description
ASGPR1 Monoclonal specifically detects ASGPR1 in Human samples. It is validated for Western Blot, Sandwich ELISA Detection.Specifications
ASGPR1 | |
Western Blot 1:20000-1:100000, Sandwich ELISA Detection 1:1000-1:10000 | |
Monoclonal | |
Purified | |
RUO | |
0.2 um filtered solution in PBS | |
432 | |
IgG | |
Protein A purified |
24 | |
Western Blot, ELISA | |
Unconjugated | |
Rabbit | |
Human | |
ASGPR, ASGPR 1, ASGP-R 1, asialoglycoprotein receptor 1, CLEC4H1ASGPR1, C-type lectin domain family 4 member H1, Hepatic lectin H1, HL-1 | |
Recombinant Human ASGPR1 Protein | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title