Learn More
ASGR2 Mouse anti-Human, Clone: OTI4B6, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF505266

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
ASGR2 is a cell surface receptor that binds to galactose-terminated glycoproteins. It transports these glycoproteins via a series of membrane vesicles and tubules to an acidic-sorting organelle where the receptor and ligand dissociates. Then the receptor is recycled back to the cell surface. There are four alternatively spliced transcript variants of this gene. This gene has multiple polyadenylation sites.Specifications
ASGR2 | |
Monoclonal | |
Unconjugated | |
ASGR2 | |
ASGP-R2, ASGPR2, CLEC4H2, HBXBP, HL-2 | |
Mouse | |
Affinity Chromatography | |
RUO | |
433 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Immunocytochemistry, Immunofluorescence, Western Blot | |
OTI4B6 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
P07307 | |
ASGR2 | |
Full length human recombit protein of human ASGR2 produced in HEK293T cell. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2b |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.