Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ASGR2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA553410
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA553410 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA553410 Supplier Invitrogen™ Supplier No. PA553410
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: YRHNYKNWAV TQPDNWHGHE LGGSEDCVEV QPDGRWNDDF CLQVYRWVCE KR Highest antigen sequence identity to the following orthologs: Mouse - 73%, Rat - 69%.

ASGR2 is a cell surface receptor that binds to galactose-terminated glycoproteins. It transports these glycoproteins via a series of membrane vesicles and tubules to an acidic-sorting organelle where the receptor and ligand dissociates. Then the receptor is recycled back to the cell surface. There are four alternatively spliced transcript variants of this gene. This gene has multiple polyadenylation sites.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ASGR2
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ASGR2
Gene Accession No. P07307
Gene Alias ANCO1; ANCO-2; ASGPR 2; ASGP-R 2; ASGPR2; ASGP-R2; Asgr; Asgr2; Asgr-2; Asialoglycoprotein receptor 2; CLEC4H2; C-type lectin domain family 4 member H2; GAC-1; HBxAg-binding protein; HBXBP; Hepatic lectin 2; hepatic lectin 2 RHL2; hepatic lectin 2, RHL2; hepatic lectin H2; Hepatic lectin R2/3; HL-2; mHL-2; Nbla00144; RHL-2
Gene Symbols ASGR2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ASGR2. Recombinant protein control fragment (Product #RP-90511).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 433
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.