Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ASH1 Antibody, DyLight 755, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB10093290IR
Description
ASH1 Polyclonal specifically detects ASH1 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
ASH1 | |
Polyclonal | |
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin | |
Absent small and homeotic disks protein 1 homolog, ASH1, ash1 (absent, small, or homeotic)-like (Drosophila), ASH1L1, ASH1-like protein, EC 2.1.1.43, FLJ10504, huASH1, KMT2HKIAA1420, Lysine N-methyltransferase 2H, probable histone-lysine N-methyltransferase ASH1L | |
Rabbit | |
Affinity Purified | |
RUO | |
55870 | |
Store at 4C in the dark. |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
DyLight 755 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
ASH1L | |
The immunogen recognized by this antibody maps to a region between residue 2919 and 2969 of human absent, small and homeotic disks protein 1 using the numbering given in entry CAI12722.1 (GeneID 55870). | |
100 μL | |
Primary | |
Human, Mouse | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction