Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ASH1 Antibody

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB10093289
Description
ASH1 Polyclonal specifically detects ASH1 in Human samples. It is validated for Immunoprecipitation, Immunohistochemistry (Negative).Specifications
ASH1 | |
Polyclonal | |
Unconjugated | |
Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide | |
Absent small and homeotic disks protein 1 homolog, ASH1, ash1 (absent, small, or homeotic)-like (Drosophila), ASH1L1, ASH1-like protein, EC 2.1.1.43, FLJ10504, huASH1, KMT2HKIAA1420, Lysine N-methyltransferase 2H, probable histone-lysine N-methyltransferase ASH1L | |
Rabbit | |
Affinity Purified | |
RUO | |
55870 | |
Human | |
IgG |
Immunoprecipitation, Immunohistochemistry | |
1.0 mg/mL | |
Immunoprecipitation 1:10-1:500, Immunohistochemistry (Negative) | |
CAI12722.1 | |
ASH1L | |
The immunogen recognized by this antibody maps to a region between residue 1925 and 1975 of human absent, small and homeotic disks protein 1 using the numbering given in entry CAI12722.1 (GeneID 55870). | |
100 μL | |
Primary | |
Human. Based on 100% sequence identity, this antibody is predicted to react with Dog, Orangutan, Chimpanzee and Northern white-cheeked gibbon. | |
Store at 4C. Do not freeze. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction