Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ASH1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5144261
Description
ASH1 Polyclonal Antibody for ICC/IF, IHC (P), ELISA
ASH1 (ASH1L) is a member of the trithorax group of transcriptional activators. The protein contains four AT hooks, a SET domain, a PHD-finger motif, and a bromodomain. It is localized to many small speckles in the nucleus, and also to cell-cell tight junctions. Diseases associated with ASH1 include Mental Retardation, Autosomal Dominant 52 and Pulpitis.
Specifications
ASH1 | |
Polyclonal | |
Unconjugated | |
ASH1L | |
8030453L17Rik; Absent small and homeotic disks protein 1 homolog; absent, small, or homeotic discs 1; Ash1; ash1 (absent, small, or homeotic)-like; ash1 (absent, small, or homeotic)-like (Drosophila); ASH1 like histone lysine methyltransferase; Ash1l; ASH1L1; ASH1-like protein; chromatin remodeling factor; E430018P19Rik; histone-lysine N-methyltransferase ASH1L; huASH1; KIAA1420; KMT2H; lysine N-methyltransferase 2H; probable histone-lysine N-methyltransferase ASH1L | |
Rabbit | |
Protein G | |
RUO | |
55870 | |
-20°C or -80°C if preferred | |
Liquid |
ELISA, Immunohistochemistry (Paraffin), Immunocytochemistry | |
3 mg/mL | |
PBS with 50% glycerol and 0.03% Proclin 300; pH 7.4 | |
Q9NR48 | |
ASH1L | |
Recombinant Human Histone-lysine N-methyltransferase ASH1L protein (1906-2160AA). | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction