Learn More
ASH2 Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
$151.31 - $568.26
Specifications
Antigen | ASH2 |
---|---|
Applications | Immunohistochemistry |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Liquid |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-157-1504
![]() |
Bethyl Laboratories, Inc
IHC00110T |
10 μL |
Each for $151.31
|
|
|||||
50-157-1503
![]() |
Bethyl Laboratories, Inc
IHC00110 |
100 μL |
Each for $568.26
|
|
|||||
Description
The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Rat, Chicken, Turkey, Bovine, Dog, Horse, Guinea pig_10141, Panda, Orangutan, Monkey, Gori
Ash2 is a component of the Set1/Ash2 histone methyltransferase (HMT) complex. This complex specifically methylates K4 of histone H3, thereby activating transcription. Methylation of K4 is blocked by premethylation of the neighboring K9, a repressor of transcription. This indicates that the Set1/Ash2 HMT complex mediates the crosstalk between K9 methylation and K4 methylation. Ash2 plays a role in hematopoiesis and may be associated with some kinds of leukemia.Specifications
ASH2 | |
Polyclonal | |
Liquid | |
RUO | |
TBS with 0.1% BSA and 0.09% sodium azide | |
ASH2, ASH2L1, ASH2L2, Bre2, ASH2-like protein, set1/Ash2 histone methyltransferase complex subunit ASH2 | |
Between 575 and C-term | |
Primary | |
4° C | |
ASH2L |
Immunohistochemistry | |
Unconjugated | |
Rabbit | |
Human, Mouse | |
Q91X20, Q9UBL3 | |
23808, 9070 | |
IgG | |
Antigen affinity chromatography | |
Antibody |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.