Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ASH2L Antibody, Alexa Fluor™ 700, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB600250AF700
Description
ASH2L Polyclonal antibody specifically detects ASH2L in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
ASH2L | |
Polyclonal | |
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin | |
ASH2, ash2 (absent, small, or homeotic)-like (Drosophila), ASH2L1ash2 (absent, small, or homeotic, Drosophila, homolog)-like, ASH2L2ASH2-like protein, Bre2, set1/Ash2 histone methyltransferase complex subunit ASH2 | |
The immunogen recognized by this antibody maps to a region between residue 50 and 100 of human Absent, Small, or Homeotic-Like 2 using the numbering given in entry NP_004665.1 (GeneID 9070). | |
0.1 mL | |
DNA Repair, Transcription Factors and Regulators | |
9070 | |
Store at 4°C in the dark. | |
IgG |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Alexa Fluor 700 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction