Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ash2L Antibody Rabbit anti-Human, Mouse, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
Supplier: MilliporeSigma ABE1972
Description
Anti-Ash2L is a highly specific rabbit polyclonal antibody that targets Set1/Ash2 histone methyltransferase complex subunit Ash2L and has been tested in Chromatin Immunoprecipitation (ChIP), Immunoprecipitation, and Western Blotting.
Specifications
Ash2L Antibody | |
Polyclonal | |
Unconjugated | |
Rabbit | |
Unpurified | |
RUO | |
Primary | |
Specificity was confirmed by decreased target band detection in lysates from Ash2L shRNA-treated C2C12 cells (Rampalli, S., et al. (2007). Nat. Struct. Mol. Biol. 14(12):1150-1156). | |
Stable for 1 year at -20°C from date of receipt.Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance. |
ChIP Assay, Immunoprecipitation, Western Blot | |
Please refer to lot specific datasheet. | |
ASH2L,ASH2L1 | |
Recombinant full-length human AshL. | |
100 μL | |
Epigenetics and Nuclear Function | |
NP_004665 | |
Human, Mouse | |
Serum |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction