Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ASH2L Monoclonal Antibody (AS 4C5), eBioscience™, Invitrogen™

Catalog No. 14506937
Encompass_Preferred
Change view
Click to view available options
Quantity:
2 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
14-506-937 2 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 14-506-937 Supplier Invitrogen™ Supplier No. 14506937
Add to Cart
Add to Cart

Rat Monoclonal Antibody

This AS 4C5 monoclonal antibody recognizes human ASH2L, which has been described as a transcriptional regulator and may play a role in hematopoiesis. AS 4C5 detects full length (unspliced) and reduced ASH2L by Western Blot and stains ASH2L in the nucleoplasm and at the plasma membrane in Immunocytochemistry. ASH2L monoclonal antibody (AS 4C5) has been validated for the following applications: Western Blot and Immunocytochemistry; AS 4C5 does not cross react with mouse ASH2L. Applications Reported: This AS 4C5 antibody has been reported for use in Western Blot and Immunocytochemistry. Applications Tested: This AS 4C5 antibody has been tested by western blot using whole cell lysates of HeLa cells (human) and NIH 3T3 or N2a (mouse cell lines) and by immunocytochemistry on HeLa cells. This AS 4C5 antibody may be used at 2 μg/mL for western blot and at 1.25 μg/mL for immunocytochemistry.

The human ASH2L gene encodes a 628 amino acid protein known as ASH2L1, or isoform 1, which contains a nuclear localization signal and PHD finger motif, suggesting that the gene product functions as a transcription regulator. Its predicted molecular weight is 68.7kDa. Alternative splicing results in possible isoforms at 60 kDa & 56.5 kDa). Human ASH2L proteins are 60% homologous to Drosophila ash2, which positively regulates expression of certain genes in early development, and contain similar, but not identical, domains, including a zinc finger motif. Homology to mouse ASH2L is 90%. ASH2L is predominantly expressed in adult heart and testis and fetal lung and liver, with barely detectable expression in adult lung, liver, kidney, prostate, and peripheral leukocytes as well as leukemia cell lines with erythroid and megakaryocytic potential, such as K562, Hel and Dami. Differentiation inducers (e.g. phorbol ester and hemin) cause different expression patterns in these cell lines, suggesting that ASH2L plays a role in hematopoiesis and is associated with particular types of leukemia. In addition ASH2L has been suggested to interact with beta-tubulin and beta-catenin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ASH2L
Applications Western Blot, Immunocytochemistry
Classification Monoclonal
Clone AS 4C5
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.2
Gene ASH2L
Gene Accession No. Q9UBL3
Gene Alias ASH 2; ASH2; ash2 (absent, small, or homeotic)-like; ash2 (absent, small, or homeotic)-like (Drosophila); ASH2 LIKE; ASH2 like histone lysine methyltransferase complex subunit; ASH2 like protein; ASH2 like, histone lysine methyltransferase complex subunit; Ash2l; ASH2L1; ASH2L2; ash2-like; ASH2-like protein; Bre 2; Bre2; set1/Ash2 histone methyltransferase complex subunit ASH2
Gene Symbols ASH2L
Host Species Rat
Purification Method Purified
Quantity 2 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9070
Target Species Human
Content And Storage 4°C, do not freeze
Product Type Antibody
Form Liquid
Isotype IgG2a κ
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.