Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ASH2L Monoclonal Antibody (OTI10D8), OriGene
SDP

Catalog No. 502543979
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-3979 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-254-3979 Supplier OriGene Supplier No. CF810986
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

ASH2L Monoclonal antibody specifically detects ASH2L in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot

Specifications

Antigen ASH2L
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI10D8
Concentration See Label
Conjugate Unconjugated
Formulation PBS with 8% trehalose and no preservative; pH 7.3
Gene AZIN1
Gene Accession No. O14977
Gene Alias ASH 2; ASH2; ash2 (absent, small, or homeotic)-like; ash2 (absent, small, or homeotic)-like (Drosophila); ASH2 LIKE; ASH2 like histone lysine methyltransferase complex subunit; ASH2 like protein; ASH2 like, histone lysine methyltransferase complex subunit; Ash2l; ASH2L1; ASH2L2; ash2-like; ASH2-like protein; Bre 2; Bre2; set1/Ash2 histone methyltransferase complex subunit ASH2
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 260-448 of human AZIN1 produced in E.coli.
Purification Method Protein A/G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51582
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Lyophilized
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.