Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ASH2L Monoclonal Antibody (OTI7D11)

Mouse Monoclonal Antibody

Supplier:  Invitrogen™ MA527381

Catalog No. PIMA527381


Only null left
Add to Cart

Description

Description

ASH2L Monoclonal Antibody for Western Blot, IHC (P)

The human ASH2L gene encodes a 628 amino acid protein known as ASH2L1, or isoform 1, which contains a nuclear localization signal and PHD finger motif, suggesting that the gene product functions as a transcription regulator. Its predicted molecular weight is 68.7kDa. Alternative splicing results in possible isoforms at 60 kDa & 56.5 kDa). Human ASH2L proteins are 60% homologous to Drosophila ash2, which positively regulates expression of certain genes in early development, and contain similar, but not identical, domains, including a zinc finger motif. Homology to mouse ASH2L is 90%. ASH2L is predominantly expressed in adult heart and testis and fetal lung and liver, with barely detectable expression in adult lung, liver, kidney, prostate, and peripheral leukocytes as well as leukemia cell lines with erythroid and megakaryocytic potential, such as K562, Hel and Dami. Differentiation inducers (e.g. phorbol ester and hemin) cause different expression patterns in these cell lines, suggesting that ASH2L plays a role in hematopoiesis and is associated with particular types of leukemia. In addition ASH2L has been suggested to interact with beta-tubulin and beta-catenin.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

ASH2L
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Q9UBL3
ASH2L
Human recombinant protein fragment corresponding to amino acids 347-628 of ASH2L produced in E.coli.
100 μL
Primary
Human
Antibody
IgG2b
Immunohistochemistry (Paraffin), Western Blot
OTI7D11
Unconjugated
ASH2L
ASH 2; ASH2; ash2 (absent, small, or homeotic)-like; ash2 (absent, small, or homeotic)-like (Drosophila); ASH2 LIKE; ASH2 like histone lysine methyltransferase complex subunit; ASH2 like protein; ASH2 like, histone lysine methyltransferase complex subunit; Ash2l; ASH2L1; ASH2L2; ash2-like; ASH2-like protein; Bre 2; Bre2; set1/Ash2 histone methyltransferase complex subunit ASH2
Mouse
Affinity Chromatography
RUO
9070
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.