Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ASH2L Mouse anti-Human, Mouse, Rat, Clone: OTI2H3, lyophilized, TrueMAB™
SDP

Catalog No. 501670392
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-167-0392 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-167-0392 Supplier OriGene Supplier No. CF810988
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO

Ash2 is a component of the Set1/Ash2 histone methyltransferase (HMT) complex. This complex specifically methylates K4 of histone H3, thereby activating transcription. Methylation of K4 is blocked by premethylation of the neighboring K9, a repressor of transcription. This indicates that the Set1/Ash2 HMT complex mediates the crosstalk between K9 methylation and K4 methylation. Ash2 plays a role in hematopoiesis and may be associated with some kinds of leukemia.

Specifications

Antigen ASH2L
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI2H3
Conjugate Unconjugated
Formulation PBS with 8% trehalose and no preservative; pH 7.3
Gene ASH2L
Gene Accession No. Q91X20, Q9UBL3
Gene Alias ASH2, ASH2L1, ASH2L2, Bre2
Gene Symbols ASH2L
Host Species Mouse
Immunogen Human recombit protein fragment corresponding to amino acids 347-628 of human ASH2L produced in E.coli.
Purification Method Affinity Chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23808, 290829, 9070
Target Species Human, Mouse, Rat
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.