Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ASH2L Polyclonal Antibody, Active Motif™
SDP

Catalog No. 501993874
Encompass
Change view
Click to view available options
Quantity:
10 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-199-3874 10 μL
50-199-3873 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-199-3874 Supplier Active Motif Supplier No. 39100
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The human ASH2L gene encodes a 628 amino acid protein known as ASH2L1, or isoform 1, which contains a nuclear localization signal and PHD finger motif, suggesting that the gene product functions as a transcription regulator. Its predicted molecular weight is 68.7kDa. Alternative splicing results in possible isoforms at 60 kDa & 56.5 kDa). Human ASH2L proteins are 60% homologous to Drosophila ash2, which positively regulates expression of certain genes in early development, and contain similar, but not identical, domains, including a zinc finger motif. Homology to mouse ASH2L is 90%. ASH2L is predominantly expressed in adult heart and testis and fetal lung and liver, with barely detectable expression in adult lung, liver, kidney, prostate, and peripheral leukocytes as well as leukemia cell lines with erythroid and megakaryocytic potential, such as K562, Hel and Dami. Differentiation inducers (e.g. phorbol ester and hemin) cause different expression patterns in these cell lines, suggesting that ASH2L plays a role in hematopoiesis and is associated with particular types of leukemia. In addition ASH2L has been suggested to interact with beta-tubulin and beta-catenin.

Specifications

Antigen ASH2L
Applications ChIP Assay, ChIP sequencing (ChIP-seq), Western Blot
Classification Polyclonal
Concentration Conc. Not Determined
Conjugate Unconjugated
Formulation whole serum with 30% glycerol and 0.035% sodium azide; pH
Gene ASH2L
Gene Accession No. Q9UBL3
Gene Alias ASH 2; ASH2; ash2 (absent, small, or homeotic)-like; ash2 (absent, small, or homeotic)-like (Drosophila); ASH2 LIKE; ASH2 like histone lysine methyltransferase complex subunit; ASH2 like protein; ASH2 like, histone lysine methyltransferase complex subunit; Ash2l; ASH2L1; ASH2L2; ash2-like; ASH2-like protein; BOS_23936; Bre 2; Bre2; LOC100338600; Set1/Ash2 histone methyltransferase complex subunit ASH2
Gene Symbols ASH2L
Host Species Rabbit
Immunogen This ASH2L antibody was raised against full-length recombinant human ASH2L protein.
Quantity 10 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9070
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.