Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ASH2L Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
$206.00 - $378.00
Specifications
Antigen | ASH2L |
---|---|
Concentration | 2 mg/ml |
Dilution | Western Blot 1:1000 |
Applications | Western Blot |
Classification | Polyclonal |
Description
ASH2L Polyclonal antibody specifically detects ASH2L in Human samples. It is validated for Western BlotSpecifications
ASH2L | |
Western Blot 1:1000 | |
Polyclonal | |
Purified | |
RUO | |
Human | |
Q9UBL3 | |
9070 | |
Primary | |
Store at -20°C short term. Aliquot and store at -80°C long term. Avoid freeze-thaw cycles. |
2 mg/ml | |
Western Blot | |
Unconjugated | |
Rabbit | |
DNA Repair, Transcription Factors and Regulators | |
PBS | |
ASH2, ash2 (absent, small, or homeotic)-like (Drosophila), ASH2L1ash2 (absent, small, or homeotic, Drosophila, homolog)-like, ASH2L2ASH2-like protein, Bre2, set1/Ash2 histone methyltransferase complex subunit ASH2 | |
Polyclonal antibody raised in rabbit against human ASH2L (absent, small, or homeotic 2), using a recombinant protein. | |
Protein G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title