Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Arlington Scientific ASI VDRL Antigen Test Kit
Shop All Arlington Scientific Products

Click to view available options
Quantity:
10 mL
Description
- Suitable for testing cerebral spinal fluid (CSF) specimens
- Easy to interpret
- Screw cap vials - no glass ampules
- Results on CAP and AAB surveys
- NCCLS procedure sheets available
Specifications
Specifications
| Content And Storage | Room Temperature |
| CE Marker | Self-declared |
| Clia Complexity | Moderate (Serum, Plasma or Whole Blood), Waived (Other) |
| Type | VDRL Test Kit |
| Disposable | Single-use |
| Includes | Two vials of VDRL Antigen, two vials of VDRL Buffered Saline, screw cap vials |
| Quantity | 10 mL |
| Form | Liquid Ready-to-Use |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction