Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Asialofetuin Antibody, Novus Biologicals™
SDP

Catalog No. NB017465 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
0.5 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB017465 0.1 mL
NB10062357 0.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB017465 Supplier Novus Biologicals Supplier No. NB100623570.1ML
Only null left
Add to Cart
Add to Cart

Sheep Polyclonal Antibody

Asialofetuin Polyclonal specifically detects Asialofetuin in Bovine samples. It is validated for Simple Western, ELISA.

Specifications

Antigen Asialofetuin
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 5.0 mg/mL
Conjugate Unconjugated
Dilution Simple Western, ELISA 1:1000-1:4000
Formulation Phosphate buffered saline with 0.09% Sodium Azide
Gene Accession No. P12763
Gene Symbols AHSG
Host Species Sheep
Immunogen Native
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Asialofetuin 100% Fetuin (ab at 1/2000) 100% Fetuin (ab at 1/8000) 50%
Target Species Bovine
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.