Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ASIC1 Monoclonal Antibody (N271/44), PE
GREENER_CHOICE

Catalog No. PIMA545595
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA545595 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIMA545595 Supplier Invitrogen™ Supplier No. MA545595
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

1 μg/mL of MA5-45595 was sufficient for detection of ASIC1 in 20 μg of rat brain lysate by colorimetric immunoblot analysis using Goat anti-mouse IgG:HRP as the secondary antibody. Detects approximately 60kDa. This antibody was formerly sold as clone S271-44.

This gene encodes a member of the degenerin/epithelial sodium channel (DEG/ENaC) superfamily. The members of this family are amiloride-sensitive sodium channels that contain intracellular N and C termini, 2 hydrophobic transmembrane regions, and a large extracellular loop, which has many cysteine residues with conserved spacing. The member encoded by this gene is expressed in most if not all brain neurons, and it may be an ion channel subunit; however, its function as an ion channel remains unknown. Cation channel with high affinity for sodium, which is gated by extracellular protons and inhibited by the diuretic amiloride. ASIC1 is permeable for Ca(2+), Li(+) and K(+) and is expressed in dorsal root ganglia, sciatic nerves, olfactory bulb, neo and allocortical regions, dentate granule cells, pyramidal cells of CA1-CA3 subfields of the hippocampal formation, habenula, basolateral amygdaloid nuclei, and in the Purkinje and granule cells of the cerebellum. It is diffusely detected over most other regions of the basal ganglia, including thalamic nuclei, substantia nigra, striatum and globus pallidus, hypothalamus, midbrain, pons, medulla and choroid plexus. Isoform 3 is expressed only in dorsal root ganglion while isoform 1 is expressed in DRG, spinal chord, trigeminal ganglia and the trigeminal mesencephalic nucleus.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ASIC1
Applications Immunohistochemistry, Western Blot, Immunocytochemistry
Classification Monoclonal
Clone N271/44
Concentration 1 mg/mL
Conjugate PE
Formulation 2.48mM MES, 95.64mM phosphate with 0.5M EDTA and no preservative; pH 7.4
Gene ASIC1
Gene Accession No. P55926, P78348, Q6NXK8
Gene Alias Accn2; ACCN2 variant 3; acid sensing (proton gated) ion channel 1; acid sensing ion channel; acid sensing ion channel 1; acid sensing ion channel subunit 1; acid-sensing (proton-gated) ion channel 1; Acid-sensing ion channel; Acid-sensing ion channel 1; acid-sensing ion channel 1a protein; AI843610; Amiloride-sensitive cation channel 2, neuronal; Asic; ASIC1; ASIC1a; B530003N02Rik; BNAC2; brain sodium channel 2; Cation channel, amiloride-sensitive, neuronal, 2; degenerin 2; hBNaC2; proton gated cation channel ASIC1
Gene Symbols ASIC1
Host Species Mouse
Immunogen Fusion protein amino acids 460-526 (Cytoplasmic C-terminus) of mouse ASIC1.
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11419, 41, 79123
Target Species Human, Mouse, Rat
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.