Learn More
Invitrogen™ ASIC1 Polyclonal Antibody

Description
Predicted to react with Mouse and Rat samples.
Specifications
Specifications
| Antigen | ASIC1 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.5% BSA, 50% glycerol and 0.02% sodium azide; pH 7.4 |
| Gene | ASIC1 |
| Gene Accession No. | P55926, P78348, Q6NXK8 |
| Gene Alias | Accn2; ACCN2 variant 3; acid sensing (proton gated) ion channel 1; acid sensing ion channel; acid sensing ion channel 1; acid sensing ion channel subunit 1; acid-sensing (proton-gated) ion channel 1; Acid-sensing ion channel; Acid-sensing ion channel 1; acid-sensing ion channel 1a protein; AI843610; Amiloride-sensitive cation channel 2, neuronal; Asic; ASIC1; ASIC1a; B530003N02Rik; BNAC2; brain sodium channel 2; Cation channel, amiloride-sensitive, neuronal, 2; degenerin 2; hBNaC2; proton gated cation channel ASIC1 |
| Gene Symbols | ASIC1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.