Learn More
Invitrogen™ ASIC2 Polyclonal Antibody

Description
This antibody is predicted to react with mouse and rat based on sequence homology.
Specifications
Specifications
| Antigen | ASIC2 |
| Applications | Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 0.3 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.09% sodium azide; pH 7.4 |
| Gene | ASIC2 |
| Gene Accession No. | Q16515 |
| Gene Alias | ACCN; ACCN1; ACIC2; acid sensing (proton gated) ion channel 2; acid sensing ion channel 2; acid sensing ion channel subunit 2; acid-sensing (proton-gated) ion channel 2; acid-sensing ion channel 2; Amiloride-sensitive brain sodium channel; amiloride-sensitive brain sodium channel 2; Amiloride-sensitive cation channel 1, neuronal; amiloride-sensitive cation channel 1, neuronal (degenerin); Amiloride-sensitive cation channel neuronal 1; Asic2; ASIC2a; Bnac1; BNaC1a; BNC1; BNC1k; Brain sodium channel 1; degenerin; hBNaC1; Mammalian degenerin homolog; MDEG; MDEG1; MDEG2; neuronal amiloride-sensitive cation channel 1 |
| Gene Symbols | ASIC2 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.