Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ASIC2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5113613
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5113613 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5113613 Supplier Invitrogen™ Supplier No. PA5113613
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LLDLLGKEED EGSHDENVST CDTMPNHSET ISHTVNVPLQ TTLGTLEEIA For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles. Predicted homology: Cow: 100%; Dog: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 86%.

ACCN1 encodes a member of the degenerin/epithelial sodium channel (DEG/ENaC) superfamily. The members of this family are amiloride-sensitive sodium channels that contain intracellular N and C termini, 2 hydrophobic transmembrane regions, and a large extracellular loop, which has many cysteine residues with conserved spacing. The member encoded by this protein may play a role in neurotransmission. In addition, a heteromeric association between this member and ACCN3 (variant 1) has been observed to co-assemble into proton-gated channels sensitive to gadolinium.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ASIC2
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene ASIC2
Gene Accession No. Q16515
Gene Alias ACCN; ACCN1; ACIC2; acid sensing (proton gated) ion channel 2; acid sensing ion channel 2; acid sensing ion channel subunit 2; acid-sensing (proton-gated) ion channel 2; acid-sensing ion channel 2; Amiloride-sensitive brain sodium channel; amiloride-sensitive brain sodium channel 2; Amiloride-sensitive cation channel 1, neuronal; amiloride-sensitive cation channel 1, neuronal (degenerin); Amiloride-sensitive cation channel neuronal 1; Asic2; ASIC2a; Bnac1; BNaC1a; BNC1; BNC1k; Brain sodium channel 1; degenerin; hBNaC1; Mammalian degenerin homolog; MDEG; MDEG1; MDEG2; neuronal amiloride-sensitive cation channel 1
Gene Symbols ASIC2
Host Species Rabbit
Immunogen Synthetic peptide directed towards the middle region of human ACCN1.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 40
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.