Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ASIC2 Rabbit anti-Human, Mouse, Rat, Polyclonal, FabGennix
Rabbit Polyclonal Antibody
Supplier: Fabgennix Inc ASIC2201AP

Description
ASIC2 Polyclonal antibody specifically detects ASIC2 in Human, Mouse, Rat samples. It is validated for ELISA, Immunoprecipitation, Western BlotSpecifications
ASIC2 | |
Polyclonal | |
Unconjugated | |
ASIC2 | |
ACCN; ACCN1; ACIC2; acid sensing (proton gated) ion channel 2; acid sensing ion channel 2; acid sensing ion channel subunit 2; acid-sensing (proton-gated) ion channel 2; acid-sensing ion channel 2; Amiloride-sensitive brain sodium channel; amiloride-sensitive brain sodium channel 2; Amiloride-sensitive cation channel 1, neuronal; amiloride-sensitive cation channel 1, neuronal (degenerin); Amiloride-sensitive cation channel neuronal 1; Asic2; ASIC2a; Bnac1; BNaC1a; BNC1; BNC1k; Brain sodium channel 1; degenerin; hBNaC1; Mammalian degenerin homolog; MDEG; MDEG1; MDEG2; neuronal amiloride-sensitive cation channel 1 | |
Synthetic peptide taken within amino acid region 450-512 on human Acid-sensing ion channel 2 protein. | |
200 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
ELISA, Immunoprecipitation, Western Blot | |
0.5-1.5 mg/mL | |
proprietary buffer with 0.5% BSA, 30% glycerol and 0.02% sodium azide; pH 7.4-7.8 | |
Q16515, Q62962, Q925H0 | |
Rabbit | |
Affinity chromatography | |
RUO | |
11418, 25364, 40 | |
-20°C | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction