Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ASIC3 Polyclonal Antibody

Catalog No. PIPA5111401
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5111401 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5111401 Supplier Invitrogen™ Supplier No. PA5111401

Rabbit Polyclonal Antibody

Immunogen sequence: CNINPLRRSR LTPNDLHWAG SALLGLDPAE HAAFLRALGR PPAPPGFMPS PTFDMAQLYA RAGHSLDDML LDCRFRGQPC GPENFTTIFT RM.

ACCN3 encodes a member of the degenerin/epithelial sodium channel (DEG/ENaC) superfamily. The members of this family are amiloride-sensitive sodium channels that contain intracellular N and C termini, two hydrophobic transmembrane regions, and a large extracellular loop, which has many cysteine residues with conserved spacing. ACCN3 encodes a member that is an acid sensor and may play an important role in the detection of lasting pH changes. In addition, a heteromeric association between this member and ACCN1 has been observed as proton-gated channels sensitive to gadolinium.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ASIC3
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.20 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ASIC3
Gene Accession No. Q9UHC3
Gene Alias ACCN3; acid sensing (proton gated) ion channel 3; acid sensing ion channel 3; acid sensing ion channel subunit 3; acid-sensing (proton-gated) ion channel 3; acid-sensing ion channel 3; Amiloride-sensitive cation channel 3; amiloride-sensitive cation channel 3, testis; Asic3; AW742291; dorsal root ASIC; DRASIC; hASIC3; hTNaC1; modulatory subunit of ASIC2a; neuronal amiloride-sensitive cation channel 3; proton gated cation channel DRASIC; proton-gated cation channel subunit; SLNAC1; Testis sodium channel 1; TNaC1
Gene Symbols ASIC3
Host Species Rabbit
Immunogen Recombinant Protein corresonding to Human ASIC3. Recombinant protein control fragment (Product #RP-108208).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9311
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.