Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ASIC3a/ASIC3 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501981966
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-1966 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-1966 Supplier Bioss Supplier No. BS12132R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ACCN3 encodes a member of the degenerin/epithelial sodium channel (DEG/ENaC) superfamily. The members of this family are amiloride-sensitive sodium channels that contain intracellular N and C termini, two hydrophobic transmembrane regions, and a large extracellular loop, which has many cysteine residues with conserved spacing. ACCN3 encodes a member that is an acid sensor and may play an important role in the detection of lasting pH changes. In addition, a heteromeric association between this member and ACCN1 has been observed as proton-gated channels sensitive to gadolinium.

Specifications

Antigen ASIC3a/ASIC3
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene ASIC3
Gene Accession No. Q9UHC3
Gene Alias Accn3; acid sensing (proton gated) ion channel 3; acid sensing ion channel 3; acid sensing ion channel subunit 3; acid-sensing (proton-gated) ion channel 3; Acid-sensing ion channel 3; Amiloride-sensitive cation channel 3; amiloride-sensitive cation channel 3, testis; ASIC3; AW742291; BOS_4613; Dorsal root ASIC; Drasic; hASIC3; hTNaC1; modulatory subunit of ASIC2a; Neuronal amiloride-sensitive cation channel 3; proton gated cation channel DRASIC; proton-gated cation channel subunit; SLNAC1; Testis sodium channel 1; TNAC1
Gene Symbols ASIC3
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human ASIC3.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9311
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.