Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ASIC4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA530649
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA530649 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA530649 Supplier Invitrogen™ Supplier No. PA530649
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: SK-N-SH, U87-MG, MCF-7. Predicted reactivity: Mouse (96%), Rat (96%), Rabbit (95%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

This gene belongs to the superfamily of acid-sensing ion channels, which are proton-gated, amiloride-sensitive sodium channels. These channels have been implicated in synaptic transmission, pain perception as well as mechanoperception. This gene is predominantly expressed in the pituitary gland, and was considered a candidate for paroxysmal dystonic choreoathetosis (PDC), a movement disorder, however, no correlation was found between mutations in this gene and PDC. Alternative splicing at this locus results in two transcript variants encoding different isoforms.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ASIC4
Applications Western Blot
Classification Polyclonal
Concentration 0.42 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7
Gene Asic4
Gene Accession No. Q96FT7
Gene Alias Accn4; acid sensing (proton gated) ion channel family member 4; acid sensing ion channel family member 4; acid sensing ion channel subunit family member 4; acid-sensing (proton-gated) ion channel family member 4; acid-sensing ion channel 4; Amiloride-sensitive cation channel 4; Amiloride-sensitive cation channel 4, pituitary; amiloride-sensitive cation channel family member 4, pituitary; ASIC4; BNAC4; brain sodium channel 4; MGC17248; MGC24860; putative acid-sensing ion channel; Spasic; Spinal chord ASIC
Gene Symbols Asic4
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the Extracellular domain of human ACCN4. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55515
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.