Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ASIC4 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5119835
This item is not returnable.
View return policy
Description
Positive Control: HepG2 cell lysate, human brain tissue lysate, K562 cell lysate, rat brain tissue, mouse pituitary tissue, SH-SY5Y. Subcellular Location: Membrane.
This gene belongs to the superfamily of acid-sensing ion channels, which are proton-gated, amiloride-sensitive sodium channels. These channels have been implicated in synaptic transmission, pain perception as well as mechanoperception. This gene is predominantly expressed in the pituitary gland, and was considered a candidate for paroxysmal dystonic choreoathetosis (PDC), a movement disorder, however, no correlation was found between mutations in this gene and PDC. Alternative splicing at this locus results in two transcript variants encoding different isoforms.
Specifications
| ASIC4 | |
| Polyclonal | |
| Unconjugated | |
| Asic4 | |
| Accn4; acid sensing (proton gated) ion channel family member 4; acid sensing ion channel family member 4; acid sensing ion channel subunit family member 4; acid-sensing (proton-gated) ion channel family member 4; acid-sensing ion channel 4; Amiloride-sensitive cation channel 4; Amiloride-sensitive cation channel 4, pituitary; amiloride-sensitive cation channel family member 4, pituitary; ASIC4; BNAC4; brain sodium channel 4; MGC17248; MGC24860; putative acid-sensing ion channel; Spasic; Spinal chord ASIC | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 241118, 55515, 63882 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| TBS with 0.2% BSA, 50% glycerol and 0.05% sodium azide; pH 7.4 | |
| Q7TNS7, Q96FT7, Q9JHS6 | |
| Asic4 | |
| Recombinant protein within human ASIC4 aa 250-450. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction