Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ASIC5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA587795
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA587795 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA587795 Supplier Invitrogen™ Supplier No. PA587795
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: HLQEITANST GSREATDFAA SHQNFSIVEF IRNKGFYLNN STLLDCEFFG KPCSPKDFAH VFTEYGNCFT FNHGETLQAK RKVSVSGRGL SLLFNVNQEA FTDNPALGFV DAGIIFVIHS PKKVPQFDGL GLLSPVGMHA RVTIRQVKTV HQEYPWGECN PNIKLQNFSS YSTSGCLKEC KAQHIKKQCG C; Positive Samples: Mouse brain, Rat brain; Cellular Location: Cell membrane, Multi-pass membrane protein.

Cation channel that gives rise to very low constitutive currents in the absence of activation. The activated channel exhibits selectivity for sodium, and is inhibited by amiloride.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ASIC5
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.37 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene ASIC5
Gene Accession No. Q9R0W5, Q9R0Y1
Gene Alias Accn5; acid sensing (proton gated) ion channel family member 5; acid sensing ion channel family member 5; acid sensing ion channel subunit family member 5; acid-sensing (proton-gated) ion channel family member 5; acid-sensing ion channel 5; Amiloride-sensitive cation channel 5; amiloride-sensitive cation channel 5, intestinal; amiloride-sensitive Na+ channel; amiloride-sensitive Na+ channel (BLINaC gene); amiloride-sensitive sodium channel; Asic5; BLINaC; brain-liver-intestine amiloride-sensitive Na(+) channel; brain-liver-intestine amiloride-sensitive sodium channel; HINaC; Human intestine Na(+) channel; Inac; inactivation no afterpotential C
Gene Symbols ASIC5
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 140-330 of human ASIC5 (NP_0591151).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 58170, 63866
Target Species Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.