Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ASK Recombinant Rabbit Monoclonal Antibody (24D8)
GREENER_CHOICE

Catalog No. FEMA550027
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEMA550027 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEMA550027 Supplier Invitrogen™ Supplier No. MA550027
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

ASK Recombinant Monoclonal Antibody for ELISA, ICC/IF, Western Blot

Activator of S phase kinase (ASK, DBF4) is a novel regulatory subunit of the human Cdc7 kinase complex that plays a pivotal role in G1/S cell cycle transition in mammalian cells. The increase in Cdc7 kinase activity at the G1/S boundary is at least partly accounted for by the elevated expression of ASK in late G1. ASK interacts with replication origins in vivo, suggesting that Cdc7 may trigger S phase by directly activating the replication initiation complexes assembled at the origins. Human ASK protein levels are regulated during the cell cycle with a pattern that matches that of human Cdc7 protein kinase activity. Human Cdc7-ASK kinase is directly involved in regulating the initiation of DNA replication by targeting MCM2 protein in mammalian cells. Nuclear localization and chromatin binding of endogenous huCdc7 and GFP-ASK is presumably imported into nuclei through its two nuclear localization signals at telophase, it may require additional signals for chromatin binding, the level of which increases at late G1 ohase. A functional link may exist between menin, a tumor suppressor protein mutated in multiple endocrine neoplasia type I (MEN1), and ASK in the regulation of cell proliferation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ASK
Applications ELISA, Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone 24D8
Concentration 0.35 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.4
Gene DBF4
Gene Accession No. Q9UBU7
Gene Alias AA545217; Activator of S phase kinase; ASK; CHIF; chiffon homolog A; Dbf4; DBF4 homolog; DBF4 homolog (S. cerevisiae); DBF4 zinc finger; DBF4 zinc finger A; DBF4A; DBF4-type zinc finger-containing protein 1; MuDBF4; Protein DBF4 homolog A; RGD1305854; ZDBF1; zinc finger, DBF-type containing 1
Gene Symbols DBF4
Host Species Rabbit
Immunogen A synthesized peptide derived from human DBF4.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10926
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.