Learn More
Promega ASK1 Kinase Enzyme System, 10microg
Catalog No. PRV3881 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
10 μg
Catalog No. Quantity
PRV3881 10 μg
1 options
Catalog No. PRV3881 Supplier Promega Supplier No. V3881
Recombinant human ASK1 (649-946) was expressed by baculovirus in Sf9 cells using an N-terminal GST tag. ASK1, also known as MAPKKK5, activates MKK3, MKK4 (SEK1) and MKK6. Overexpression of ASK1 induces apoptotic cell death.
  • Easily Screen and Profile ASK1 Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP−Glo™ Assay for bioluminescent detection of kinase activity
Specifications
Quantity 10 μg
Content And Storage <-65°C