Learn More
Promega ASK1 Kinase Enzyme System, 1mg
Catalog No. PRV3882 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
1 mg
Catalog No. Quantity
PRV3882 1 mg
1 options
Catalog No. PRV3882 Supplier Promega Supplier No. V3882
Add to Cart
Add to Cart
Request a Quote
Recombinant human ASK1 (649-946) was expressed by baculovirus in Sf9 cells using an N-terminal GST tag. ASK1, also known as MAPKKK5, activates MKK3, MKK4 (SEK1) and MKK6. Overexpression of ASK1 induces apoptotic cell death.
  • Easily Screen and Profile ASK1 Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP−Glo™ Assay for bioluminescent detection of kinase activity
Specifications
Quantity 1 mg
Content And Storage <-65°C