Learn More
ASL Mouse anti-Human, Clone: OTI9G2, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF804902

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
ASL encodes a member of the lyase 1 family. The encoded protein forms a cytosolic homotetramer and primarily catalyzes the reversible hydrolytic cleavage of argininosuccinate into arginine and fumarate, an essential step in the liver in detoxifying ammonia via the urea cycle. Mutations in this gene result in the autosomal recessive disorder argininosuccinic aciduria, or argininosuccinic acid lyase deficiency. A nontranscribed pseudogene is also located on the long arm of chromosome 22. Alternatively spliced transcript variants encoding different isoforms have been described.Specifications
ASL | |
Monoclonal | |
Unconjugated | |
ASL | |
ASAL | |
Mouse | |
Affinity Chromatography | |
RUO | |
435 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Immunohistochemistry (Paraffin), Western Blot | |
OTI9G2 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
P04424 | |
ASL | |
Full length human recombit protein of human ASL produced in HEK293T cell. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.