Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ASL Polyclonal Antibody
Supplier: Thermo Scientific PA5117795
This item is not returnable.
View return policy
Description

Specifications
ASL | |
Polyclonal | |
Unconjugated | |
ASL | |
2510006M18Rik; argininosuccinase; argininosuccinate lyase; Arginosuccinase; ASAL; Asl | |
Rabbit | |
Antigen Affinity Chromatography, Protein A | |
RUO | |
435 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
1.0 mg/mL | |
PBS with 0.03% ProClin 300; pH 7 | |
P04424 | |
ASL | |
E. coli-derived Human Argininosuccinate lyase fragment. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction