Learn More
Invitrogen™ ASMT Polyclonal Antibody

Description
Peptide sequence: GSSEDQAYRL LNDYANGFMV SQVLFAACEL GVFDLLAEAP GPLDVAAVAA Sequence homology: Human: 100%.
Specifications
Specifications
| Antigen | ASMT |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 2% sucrose and 0.09% sodium azide |
| Gene | ASMT |
| Gene Accession No. | P46597 |
| Gene Alias | acetylserotonin methyltransferase (Y chromosome); acetylserotonin N-methyltransferase; acetylserotonin O-methyltransferase; ASMT; ASMTY; Hiomt; HIOMTY; hydroxyindole O-methyltransfease; hydroxyindole O-methyltransferase; hydroxyindole-O-methyltransferase |
| Gene Symbols | ASMT |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.