Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ASMT Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501987627
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-7627 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-7627 Supplier Bioss Supplier No. BS6961R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ASMT belongs to the methyltransferase superfamily, and is located in the pseudoautosomal region (PAR) at the end of the short arms of the X and Y chromosomes. The encoded enzyme catalyzes the final reaction in the synthesis of melatonin, and is abundant in the pineal gland.

Specifications

Antigen ASMT
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene ASMT
Gene Accession No. P46597
Gene Alias acetylserotonin methyltransferase (Y chromosome); acetylserotonin N-methyltransferase; Acetylserotonin O-methyltransferase; ASMT; ASMTY; HIOMT; HIOMTY; Hydroxyindole O-methyltransferase
Gene Symbols ASMT
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human Acetylserotonin O-methyltransferase (ASMT).
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 438
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.