Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ASMTL Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA531731
Description
Recommended positive controls: HepG2, HCT116. Predicted reactivity: Chimpanzee (96%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
The protein encoded by this gene has an N-terminus that is similar to the multicopy associated filamentation (maf) protein of Bacillus subtilis and to orfE of Escherichia coli, while the C-terminus is similar to N-acetylserotonin O-methyltransferase. This gene is located in the pseudoautosomal region 1 (PAR1) of X and Y chromosomes. Three transcript variants encoding different isoforms have been found for this gene.
Specifications
ASMTL | |
Polyclonal | |
Unconjugated | |
ASMTL | |
acetylserotonin N-methyltransferase-like; acetylserotonin O-methyltransferase-like; ASMTL; ASMTLX; ASMTLY; ASTML; dTTP/UTP pyrophosphatase; dTTPase/UTPase; N-acetylserotonin O-methyltransferase-like protein; Nucleoside triphosphate pyrophosphatase; Nucleotide PPase; Nucleotide pyrophosphatase; Probable bifunctional dTTP/UTP pyrophosphatase/methyltransferase protein | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
8623 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
O95671 | |
ASMTL | |
Recombinant fragment corresponding to a region within amino acids 95 and 328 of Human ASMTL. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction