Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ASMTL Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA554530
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA554530 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA554530 Supplier Invitrogen™ Supplier No. PA554530
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: RLLDICAAMG LLEKTEQGYS NTETANVYLA SDGEYSLHGF IMHNNDLTWN LFTYLEFAIR EGTNQHHRAL GKKAEDLFQD AYYQSPETRL RFMRAMHGMT KLTACQVATA FNLSRFSSAC DVGGCTGALA RELAREYPRM QVTV Highest antigen sequence identity to the following orthologs: Mouse - 38%, Rat - 40%.

The protein encoded by this gene has an N-terminus that is similar to the multicopy associated filamentation (maf) protein of Bacillus subtilis and to orfE of Escherichia coli, while the C-terminus is similar to N-acetylserotonin O-methyltransferase. This gene is located in the pseudoautosomal region 1 (PAR1) of X and Y chromosomes. Three transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ASMTL
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ASMTL
Gene Accession No. O95671
Gene Alias acetylserotonin N-methyltransferase-like; acetylserotonin O-methyltransferase-like; ASMTL; ASMTLX; ASMTLY; ASTML; dTTP/UTP pyrophosphatase; dTTPase/UTPase; N-acetylserotonin O-methyltransferase-like protein; Nucleoside triphosphate pyrophosphatase; Nucleotide PPase; Nucleotide pyrophosphatase; Probable bifunctional dTTP/UTP pyrophosphatase/methyltransferase protein
Gene Symbols ASMTL
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ASMTL. Recombinant protein control fragment (Product #RP-92232).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8623
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.