Learn More
Invitrogen™ ASMTL Polyclonal Antibody

Description
Positive test controls include: MCF7, 22Rv1, SW620, NIH/3T3, HeLa, Mouse pancreas, Mouse thymus, Mouse testis. Immunogen sequence: MVLCPVIGKL LHKRVVLASA SPRRQEILSN AGLRFEVVPS KFKEKLDKAS FATPYGYAME TAKQKALEVA NRLYQKDLRA PDVVIGADTI VTVGGLILEK PVDKQDAYRM LSRLSGREHS VFTGVAIVHC SSKDHQLDTR VSEFYEETKV KFSELSEELL WEYVHSGEPM DKAGGYGIQA LGGMLVESVH GDFLNVVGFP LNHFCKQLVK LYYPPRPEDL RRSVKHDSIP AADTFEDLSD VEGGGSEPTQ RDAGSRDEKA EAGEAGQATA EAECHRTRET LPPFPTRLLE.
Specifications
Specifications
| Antigen | ASMTL |
| Applications | ELISA, Immunoprecipitation, Western Blot |
| Classification | Polyclonal |
| Concentration | 0.23 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | ASMTL |
| Gene Accession No. | O95671 |
| Gene Alias | acetylserotonin N-methyltransferase-like; acetylserotonin O-methyltransferase-like; ASMTL; ASMTLX; ASMTLY; ASTML; dTTP/UTP pyrophosphatase; dTTPase/UTPase; N-acetylserotonin O-methyltransferase-like protein; Nucleoside triphosphate pyrophosphatase; Nucleotide PPase; Nucleotide pyrophosphatase; Probable bifunctional dTTP/UTP pyrophosphatase/methyltransferase protein |
| Gene Symbols | ASMTL |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.