Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ASNA1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP179797
Description
ASNA1 Polyclonal specifically detects ASNA1 in Human samples. It is validated for Western Blot.Specifications
ASNA1 | |
Polyclonal | |
Unconjugated | |
PBS, 2% Sucrose with 0.09% Sodium Azide | |
ARSA, arsA arsenite transporter, ATP-binding, homolog 1 (bacterial), ARSA1, ARSA-I, Arsenical pump-driving ATPase, Arsenite-stimulated ATPase, ATPase ASNA1, EC 3.6, EC 3.6.3.16, GET3, golgi to ER traffic 3 homolog, hARSA-I, hASNA-I, MGC3821, Transmembrane domain recognition complex 40 kDa ATPase subunit, transmembrane domain recognition complex, 40kDa, TRC40arsA (bacterial) arsenite transporter, ATP-binding, homolog 1 | |
Rabbit | |
39 kDa | |
100 μL | |
metabolism | |
439 | |
Human, Mouse, Rat, Bovine, Canine, Equine, Guinea Pig, Rabbit, Zebrafish | |
IgG |
Western Blot | |
0.5 mg/ml | |
Western Blot 1.0 ug/ml | |
NP_004308 | |
GET3 | |
Synthetic peptide directed towards the C terminal of human ASNA1The immunogen for this antibody is ASNA1. Peptide sequence: EARHKIQAKYLDQMEDLYEDFHIVKLPLLPHEVRGADKVNTFSALLLEPY | |
Affinity purified | |
RUO | |
Primary | |
Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction