Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ASNA1 Polyclonal Antibody
ASNA1 Polyclonal Antibody
Supplier: Thermo Scientific PA545067
Description
ASNA1 Polyclonal Antibody for Western Blot

Specifications
ASNA1 | |
Polyclonal | |
Unconjugated | |
GET3 | |
1810048H22Rik; ARSA; arsA arsenite transporter, ATP-binding, homolog 1; arsA arsenite transporter, ATP-binding, homolog 1 (bacterial); ARSA1; ARSA-I; Arsenical pump-driving ATPase; Arsenical resistance ATPase; arsenite-stimulated ATPase; arsenite-translocating ATPase; Arsenite-transporting ATPase; asna1; ASNA-I; ATPase asna1; ATPase GET3; GET3; guided entry of tail-anchored proteins factor 3, ATPase; hARSA-I; hASNA-I; MGC3821; si:dkey-121b10.2; Transmembrane domain recognition complex 40 kDa ATPase subunit; TRC40; wu:fd05d03; zgc:56540; zgc:56540 protein; zgc:86799 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
439 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
O43681 | |
GET3 | |
Synthetic peptide directed towards the C-terminal of human ASNA1 (aa 293-342). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction