Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ASNA1 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501988171
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-8171 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-8171 Supplier Bioss Supplier No. BS7972R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ASNA1 is the human homolog of the bacterial arsA gene. In E. coli, ArsA ATPase is the catalytic component of a multisubunit oxyanion pump that is responsible for resistance to arsenicals and antimonials.

Specifications

Antigen ASNA1
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene GET3
Gene Accession No. O43681
Gene Alias 1810048H22Rik; ARR4; ARSA; arsA arsenite transporter, ATP-binding, homolog 1; arsA arsenite transporter, ATP-binding, homolog 1 (bacterial); ARSA1; ARSA-I; Arsenical pump-driving ATPase; Arsenical resistance ATPase; Arsenite-stimulated ATPase; Arsenite-translocating ATPase; Arsenite-transporting ATPase; asna1; ASNA-I; ATPase ASNA1; ATPase GET3; BOS_7427; D2371; GET3; Golgi to ER traffic protein 3; guanine nucleotide exchange factor GET3; Guided entry of tail-anchored proteins 3; hARSA-I; hASNA-I; MGC3821; si:dkey-121b10.2; Transmembrane domain recognition complex 40 kDa ATPase subunit; TRC40; wu:fd05d03; YDL100C; zgc:56540; zgc:56540 protein; zgc:86799
Gene Symbols GET3
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human ASNA1.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 439
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.