Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Asparagine Synthetase Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5109421
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5109421 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5109421 Supplier Invitrogen™ Supplier No. PA5109421
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LHALYDNVEK LFPGFEIETV KNNLRILFNN AVKKRLMTDR RIGCLLSGGL DSSLVAATLL KQLKEAQVQY PLQTFAIGME DSPDLLAARK VADHIGSEHY EVLFNSEEGI QALDEVIFSL ETYDITTVRA SVGMYLISKY IRKNTDSVVI FSGEGSDELT QGYIYFHKAP SPEKAEEESE RLLRELYLFD VLRADRTTAA HGLELRVPFL DHRFSSYYLS LPPEMRIPKN GIEKHLLRET FEDSNLIPKE ILWRPKEAFS DGITSVKNSW FKILQEYVEH QVDDAMMANA AQKFPFNTPK TKEGYYYRQV FERHYPGRAD WLSHYWMPKW INATDPSART LTHYKSAVKA.

ASNS (asparagine synthetase [glutamine-hydrolyzing]) is involved in the subpathway that synthesizes f L-asparagine from L-aspartate. Mutations affecting the gene can result in asparagine synthetase deficiency (ASNSD).
TRUSTED_SUSTAINABILITY

Specifications

Antigen Asparagine Synthetase
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.71 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene Asns
Gene Accession No. P08243, P49088, Q61024
Gene Alias ASNS; ASNSD; asparagine synthetase; asparagine synthetase (glutamine-hydrolyzing); asparagine synthetase [glutamine-hydrolyzing]; Cell cycle control protein TS11; Glutamine-dependent asparagine synthetase; TS11; TS11 cell cycle control protein
Gene Symbols Asns
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 212-561 of human ASNS (NP_0016643).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 25612, 27053, 440
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.