Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sure-Vue™ ASO Latex Test Kit
SDP

Catalog No. 23038107 Shop All Fisherbrand Products
Click to view available options
Quantity:
100 Tests
50 Tests
500 Tests
This item is not returnable. View return policy
This item is not returnable. View return policy

The Sure-Vue ASO Latex Slide Test is a convenient and specific latex slide agglutination assay for the qualitative and semiquantitative detection of Antistreptolysin-O (ASO) antibodies in human serum.

  • The ASO Slide Test is a latex slide agglutination assay for the qualitative and semiquantitative detection of antistreptolysin-O (ASO) antibodies in human serum.
  • No initial dilution of patient samples is required for this test.
  • These materials are intended to be acquired, possessed and used only by health professionals.

Specifications

Content And Storage 2°C to 8°C
Control Sets Yes
CPT Code 86060
CE Marker Self-declared
Clia Complexity Moderate
Detectable Analytes Anti-Streptolysin O (ASO)
Detection Method Agglutination
Sample Type Human Serum
Sensitivity 200 IU/mL
Disposable Single-use
Includes Latex reagent, controls, cards, and applicators
Quantity 500 Tests
Sample Size 30 μL
Specificity >95%
State at Room Temperature Liquid
Sterility Non-sterile
Test Time 2 Minutes
Form Ready-to-Use (RTU)
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.