Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ ASP 7663
GREENER_CHOICESDP

Catalog No. 517850 Shop All Novus Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
517850 50 mg
517810 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 517850 Supplier R&D Systems™ Supplier No. 5178/50

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Selective TRPA1 activator

ASP 7663 is a selective TRPA1 activator (EC50 values are ∽0.5 μM in human, mouse and rat). Displays little or no affinity at more than 60 other related receptors, channels and enzymes. Stimulates 5-HT release from QGP-1 cells. Exhibits an abdominal analgesic effect in vivo. Orally bioavailable.

Specifications

Chemical Name or Material ASP 7663
CAS 1190217-35-6
Synonym (2E)-2-[7-Fluoro-1,2-dihydro-1-(2-methylpropyl)-2-oxo-3H-indol-3-ylidene]acetic acid
Quantity 50 mg
Solubility Soluble to 100mM in DMSO and to 100mM in ethanol
Product Line Tocris
Molecular Formula C14H14FNO3
Formula Weight 263.26
Physical Form Solid
Product Type Bioactive Small Molecule
Purity >98%
Show More Show Less

Research Use Only. Not For Human or Veterinary Use

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.